Learn More
Description
- A convenient method to label proteins with varying amounts maleimide for crosslinking or conjugation with a thiol/sulfhydryl containing moiety
- Sulfo-SMCC contained in this kit readily reacts with the primary amines on a protein to form a covalently attached maleimide group which can be further reacted with a reactive thiol or free cystine to create diverse bioconjugates
- Thermo Scientific™ Sulfo-SMCC is a heterobifunctional crosslinker which contains a sulfonated N-hydroxysuccinimide (NHS) ester and maleimide group that allow covalent conjugation of amine- and thiol/sulfhydryl-containing molecules
- Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins
- This kit contains sufficient reagents for five labeling reactions of up to 100 μL sample with a concentration of 1-10 mg/mL
- Sulfo-SMCC reagent readily dissolves in water
Specifications
Specifications
| Quantity | 1 Kit |
| Product Type | Protein Labeling Kit |
| Content And Storage | Contains 5 vials of sulfo-SMCC, 5 Zeba Dye & Biotin Removal columns, and BupH™ Phosphate Buffered Saline pack. Store sulfo-SMCC desiccated at –20°C, Zeba Dye and Biotin Removal columns at 2–8°C. |
| Chemical Reactivity | Amine-to-sulfhydryl |
| Labeling Method | Chemical Labeling |
| Shipping Condition | Wet Ice |
| Reactive Moiety | Sulfo-NHS, Maleimide |
| Product Line | EZ-Link |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.