Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ NHS-LC-LC-Biotin

Catalog No. PI21343 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21343 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21343 Supplier Thermo Scientific™ Supplier No. 21343
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylate protein amines with this doubly-long-chain (30.5A), NHS-ester biotinylation reagent containing a membrane permeable spacer arm for intracellular labeling.

Thermo Scientific™ EZ-Link NHS-LC-LC-Biotin is a doubly long-chain, NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for intracellular labeling.

Highlights:

  • Longest spacer arm available to reduce steric hindrance when binding several biotinylated molecules to one avidin complex
  • Less water-soluble than sulfonated form
  • Dissolve in organic solvent (DMSO or DMF) before use
TRUSTED_SUSTAINABILITY

Specifications

Product Type NHS-LC-LC-Biotin
Quantity 50 mg
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Long
Label Type Biotin & Analogs
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.