Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Micro NHS-PEG4-Biotinylation Kit

Catalog No. pi21955
Change view
Click to view available options
Quantity:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21955 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21955 Supplier Thermo Scientific™ Supplier No. 21955
Only null left
Add to Cart
Edge
Add to Cart

Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Micro NHS-PEG4-Biotinylation Kit
Product Type Micro NHS-PEG4-Biotinylation Kit
Quantity 8 reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link NHS-PEG4-Biotin, 8 x 2 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store microtubes of biotin reagent at -20°C. Store PBS at room temperature. Store desalting column at 4°C. Shipped at ambient temperature.
Labeling Scale 100 μg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Spacer Pegylated
Labeling Target Antibodies, Proteins
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.