Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Pentylamine-Biotin

Catalog No. PI21345 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21345 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21345 Supplier Thermo Scientific™ Supplier No. 21345
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (18.9A) biotinylation reagent.

Thermo Scientific™ EZ-Link Pentylamine-Biotin is biotin that has been conjugated to pentylenediamine to add a short, amine-terminated spacer to its valeric acid group, providing a simple amine-modified compound for different applications.

Highlights:

  • Requires the use of EDC crosslinking agent to couple to carboxylic acid groups or 5' phosphate groups
  • Relatively water-soluble
  • Forms an amide bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pentylamine-Biotin
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine
Solubility Water
Label or Dye Biotin
Spacer Short
Label Type Biotin & Analogs
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.