Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ PFP-Biotin

Catalog No. pi21218
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21218 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21218 Supplier Thermo Scientific™ Supplier No. 21218

Biotinylate primary amines in proteins as well as secondary amines in DNA or RNA with this very short (9.6A) pentafluorophenyl-ester biotinylation reagent.

Thermo Scientific™ EZ-Link PFP-biotin is a pentafluorophenyl-ester biotinylation reagents that has similar properties to NHS-biotin but is more reactive and will target both primary and secondary amines for biotinylation.

Highlights:

  • Reacts with primary and secondary amines at pH 7-9
  • Pentafluorophenyl esters are more reactive than NHS-esters
  • Easier to handle and store than NHS-esters
  • Creates a stable amide bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type PFP-Biotin
Quantity 50 mg
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, TFP Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Short
Label Type Biotin & Analogs
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.