Learn More
OriGene EZ-LNP RNA Transfection Kit - 1.0 mL
SDP
Catalog No. 5000600186
Encompass
Instrument-free LNP kit enabling effective RNA transfection in vitro and RNA delivery in vivo.Components: 4 vial (1 LNP + 3 buffer)- EZ-LNP, 1 mL- RNA Buffer, 1 mL (x3)Stability: 6 monthsStorage: 4°C for short term (1-2 months); -20°C for long-term. RNA buffer can be stored at 4°C for both short and long-term storage.
Catalog No. 50-006-00186 Supplier OriGene Supplier No. TT300004L
Instrument-free LNP kit enabling effective RNA transfection in vitro and RNA delivery in vivo.Components: 4 vial (1 LNP + 3 buffer)- EZ-LNP, 1 mL- RNA Buffer, 1 mL (x3)Stability: 6 monthsStorage: 4°C for short term (1-2 months); -20°C for long-term. RNA buffer can be stored at 4°C for both short and long-term storage.