Learn More
OriGene EZ-LNP Spleen Delivery Kit - 1.0 mL
SDP
Catalog No. 5000600187
Encompass
Instrument-free LNP kit supporting efficient targeted delivery to spleen cells for in vivo applications.Components: 4 vial (1 LNP + 3 buffer)- EZ-LNP, 1 mL- RNA Buffer, 1 mL (x3)Stability: 6 monthsStorage: 4°C for short term (1-2 months); -20°C for long-term. RNA buffer can be stored at 4°C for both short and long-term storage.
Catalog No. 50-006-00187 Supplier OriGene Supplier No. TT300006
Instrument-free LNP kit supporting efficient targeted delivery to spleen cells for in vivo applications.Components: 4 vial (1 LNP + 3 buffer)- EZ-LNP, 1 mL- RNA Buffer, 1 mL (x3)Stability: 6 monthsStorage: 4°C for short term (1-2 months); -20°C for long-term. RNA buffer can be stored at 4°C for both short and long-term storage.