Learn More
Bio Rad Laboratories EZ LOAD 100 BP PCR MOLECULAR R
Catalog No. 509012806
Encompass
Ruler, Molecular; Bio-Rad; EZ Load; Ready for direct loading; No contaminating plasmid DNA; 500uL; 100 to 3000 bp; Includes 1mL 5x nucleic acid sample buffer; 100 applications
Catalog No. 50-901-2806 Supplier Bio Rad Laboratories Supplier No. 1708353
Ruler, Molecular; Bio-Rad; EZ Load; Ready for direct loading; No contaminating plasmid DNA; 500uL; 100 to 3000 bp; Includes 1mL 5x nucleic acid sample buffer; 100 applications