Learn More
Apexbio Technology LLC EZ Cap Mouse CD252(OX40L) mRNA (m1Ψ HA tag) 100ug (1mg/mL)
SDP
Supplier:  Apexbio Technology LLC R10780.1
Encompass_Preferred
Mouse CD252 mRNA encodes cytokine OX40 ligand (OX40L) expressed on the surface of activated antigen-presenting cells such as dendritic cells B cells and macrophages Binding to the OX40 receptor (CD134) the CD252-OX40 signaling pathway plays a key role in the immune system promoting T cell proliferation survival and effector functions This signaling pathway significantly influences immune response regulation immune tolerance maintenance and anti-tumor immunity CD252 is involved in the pathological processes of autoimmune diseases and chronic inflammation EZ Cap Mouse CD252 (OX40L) mRNA (m1 HA tag) is provided at 1 mg/mL It expresses mouse-derived CD137 cytokine tagged with HA facilitating subsequent detection It contains a cap 1 structure generated via co-transcriptional capping Incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7473