Learn More
Apexbio Technology LLC EZ Cap Mouse CD86 mRNA (m1Ψ HA tag) 5x1mg (1mg/mL)
SDP
Supplier:  Apexbio Technology LLC R10765
Encompass_Preferred
Mouse CD86 mRNA encodes CD86P primarily expressed on antigen-presenting cells It binds to CD28 and CTLA-4 receptors on T cells to regulate T-cell activation and immune responses The interaction between CD86 and CD28 promotes T-cell activation and proliferation whereas its binding to CTLA-4 inhibits T-cell activity maintaining immune system balance Expression and function of CD86 are critical in various immune responses including infections tumor immunity and autoimmune diseases CD86 activity may offer novel therapeutic strategies for various immune-related disorders EZ Cap Mouse CD86 mRNA (m1 HA tag) is provided at 1 mg/mL expressing mouse-derived CD86 cytokine fused with HA tag facilitating subsequent detection It contains a cap 1 structure generated via co-transcriptional capping Incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7433