Learn More
Apexbio Technology LLC EZ Cap Mouse RLI mRNA (m1Ψ) 1mg (1mg/mL)
SDP
Supplier:  Apexbio Technology LLC R10751
Encompass_Preferred
Mouse RLI mRNA encodes a fusion protein composed of interleukin-15 (IL-15) fused to the sushi domain of its receptor IL-15R representing an innovative immunotherapeutic agent As a crucial cytokine IL-15 stimulates proliferation of natural killer (NK) cells and T cells thus enhancing immune responses against tumor cells and viruses The IL-15/IL-15R -sushi fusion protein shows promise in cancer immunotherapy by enhancing immune cell function and promoting clearance of tumor cells EZ Cap Mouse RLI mRNA (m1 )is supplied at 1 mg/mL expresses mouse-derived RLI fusion protein It contains a cap 1 structure generated through co-transcriptional capping Incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7314