Learn More
Apexbio Technology LLC EZ Cap Mouse IL-12 mRNA (m1Ψ) 5x1mg (1mg/mL)
SDP
Catalog No. 503107391
Encompass_Preferred
Mouse IL-12 mRNA encodes a cytokine Interleukin-12 secreted by antigen-presenting cells such as dendritic cells macrophages and B cells It promotes the activation and proliferation of T cells and natural killer (NK) cells thus enhancing the body s immune response IL-12 induces the differentiation of T cells into Th1 cells thereby enhancing cell-mediated immunity and increasing the production of interferon- (IFN- ) crucial for antiviral and antitumor immune responses EZ Cap Mouse IL-12 mRNA (m1 ) is provided at 1 mg/mL encoding mouse IL-12 cytokine It contains a cap 1 structure generated through co-transcriptional capping Incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7391 Supplier Apexbio Technology LLC Supplier No. R10585
Mouse IL-12 mRNA encodes a cytokine Interleukin-12 secreted by antigen-presenting cells such as dendritic cells macrophages and B cells It promotes the activation and proliferation of T cells and natural killer (NK) cells thus enhancing the body s immune response IL-12 induces the differentiation of T cells into Th1 cells thereby enhancing cell-mediated immunity and increasing the production of interferon- (IFN- ) crucial for antiviral and antitumor immune responses EZ Cap Mouse IL-12 mRNA (m1 ) is provided at 1 mg/mL encoding mouse IL-12 cytokine It contains a cap 1 structure generated through co-transcriptional capping Incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo