Learn More
Apexbio Technology LLC EZ Cap Mouse IL-36γ mRNA (m1Ψ HA tag) 5x1mg (1mg/mL)
SDP
Supplier:  Apexbio Technology LLC R10745
Encompass_Preferred
Mouse IL-36 mRNA encodes cytokine interleukin-36 (IL-36)Gamma primarily involved in inflammatory responses and immune regulation IL-36 acts through binding to the IL-36 receptor activating downstream signaling pathways like NF- B and MAPK promoting the release of inflammatory cytokines and activation of immune cells IL-36 is involved in multiple inflammatory disorders including psoriasis inflammatory bowel diseases and rheumatoid arthritis IL-36 may have pro-inflammatory functions in the pathological processes of these diseases making it a potential therapeutic target EZ Cap Mouse IL-36 mRNA (m1 HA tag) is supplied at 1 mg/mL It expresses mouse IL-36 cytokine fused with an HA tag facilitating subsequent detection With cap 1 structure generated through co-transcriptional capping incorporation of modified nucleotides m1 along with a poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7318