Learn More
MSC Rochester Midland Corporation Proportioner, Foamer, Regulator Knob, Hose Inlet, Portable
Catalog No. 502839907
Encompass
The Rochester Midland Corporation portable foamer proportioner is designed for efficient dispensing of solutions, particularly for EZ-Mix products. It features a regulator knob for precise adjustment and a hose inlet for convenient connection, offering portability and ease of use in various applications.
  • Portable design for flexible use.
  • Easy to use for quick setup and operation.
  • Efficient dispensing of solutions.
  • Regulator knob allows for precise adjustment.
  • Hose inlet for convenient connection.
  • Constructed with durable plastic body material.
  • Offers high flow and low flow mix ratio options.
  • Compatible with various bottles.
  • Operates within a minimum inlet pressure of 25 psi.
  • Withstands a maximum pressure of 85 psi.
  • Designed for use with EZ-Mix products.
Catalog No. 50-283-9907 Supplier MSC Supplier No. 90709312
The Rochester Midland Corporation portable foamer proportioner is designed for efficient dispensing of solutions, particularly for EZ-Mix products. It features a regulator knob for precise adjustment and a hose inlet for convenient connection, offering portability and ease of use in various applications.
  • Portable design for flexible use.
  • Easy to use for quick setup and operation.
  • Efficient dispensing of solutions.
  • Regulator knob allows for precise adjustment.
  • Hose inlet for convenient connection.
  • Constructed with durable plastic body material.
  • Offers high flow and low flow mix ratio options.
  • Compatible with various bottles.
  • Operates within a minimum inlet pressure of 25 psi.
  • Withstands a maximum pressure of 85 psi.
  • Designed for use with EZ-Mix products.