Learn More
NRD LLC EZ MOUNT FOR 2U500
SDP
Catalog No. NC0936327
Encompass_Preferred
E_Z MOUNT FOR STATICMASTER 2U500 IONIZER - HOLDER FOR DEVICE INSIDE BALANCE CHAMBER
Catalog No. NC0936327 Supplier NRD LLC Supplier No. BF3
E_Z MOUNT FOR STATICMASTER 2U500 IONIZER - HOLDER FOR DEVICE INSIDE BALANCE CHAMBER