Learn More
Anatech Ltd EZ Mucicarmine 1qt
Catalog No. NC1374648
Encompass_Preferred
EZ Mucicarmine is made of Aluminum Chloride (cas#7446-70-0), Carmine (cas#1390-65-4), and Thymol (cas# 89-83-8). This product comes in 1qt.
Catalog No. NC1374648 Supplier Anatech Ltd Supplier No. 871
EZ Mucicarmine is made of Aluminum Chloride (cas#7446-70-0), Carmine (cas#1390-65-4), and Thymol (cas# 89-83-8). This product comes in 1qt.