Learn More
Omega Biotek Inc E.Z.N.A. Viral RNA Kit
Catalog No. NC1699186
Encompass
The E.Z.N.A. Viral RNA Kit is designed for the isolation of viral DNA and viral RNA from cell-free fluids such as plasma, serum, urine and cell culture supernatant. The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues and bacteria. RNA purified using the E.Z.N.A. Viral RNA method is ready for all downstream applications such as RT-PCR.
Catalog No. NC1699186 Supplier Omega Biotek Inc Supplier No. R687402
The E.Z.N.A. Viral RNA Kit is designed for the isolation of viral DNA and viral RNA from cell-free fluids such as plasma, serum, urine and cell culture supernatant. The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues and bacteria. RNA purified using the E.Z.N.A. Viral RNA method is ready for all downstream applications such as RT-PCR.