Learn More
Zymo Research Corporation EZ Nucleosomal DNA Prep Kit, 20 preps
SDP
Catalog No. 50444364
Encompass_Preferred
Atlantis dsDNase is a double-strand DNA specific endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5’- phosphate and 3’-hydroxyl termini. Atlantis dsDNase digestion yields very homogeneous populations of core nucleosomes and purification of the nucleosome-associated DNA is performed using Zymo Research’s proven Fast-Spin column technology. The result is pure nucleosomal DNA ready for analysis in less than 45 minutes.
Catalog No. 50-444-364 Supplier Zymo Research Corporation Supplier No. D5220
Atlantis dsDNase is a double-strand DNA specific endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5’- phosphate and 3’-hydroxyl termini. Atlantis dsDNase digestion yields very homogeneous populations of core nucleosomes and purification of the nucleosome-associated DNA is performed using Zymo Research’s proven Fast-Spin column technology. The result is pure nucleosomal DNA ready for analysis in less than 45 minutes.