Learn More
Apexbio Technology LLC EZ Cap OVA mRNA 100ug (1mg/mL)
SDP
Catalog No. 503107035
Encompass_Preferred
Ovalbumin (OVA) mRNA encodes a egg white glycoprotein with a molecular weight of approximately 45 000 daltons It can induce moderate immunogenicity in organisms and is commonly used as an immunogen in immunological experiments such as establishing animal models for hypersensitivity asthma etc EZ Cap OVA mRNA is provided at 1 mg/mL capped by co-transcriptional capping with a Cap 1 structure Compared to Cap 0 structures (such as ARCA and mCap) Cap 1-containing mRNA is more suitable for mammalian systems and exhibits higher transcription efficiency Additionally OVA mRNA is polyadenylated reducing host cell immune responses and enhancing mRNA stability and translational efficiency
Catalog No. 50-310-7035 Supplier Apexbio Technology LLC Supplier No. R10270.1
Ovalbumin (OVA) mRNA encodes a egg white glycoprotein with a molecular weight of approximately 45 000 daltons It can induce moderate immunogenicity in organisms and is commonly used as an immunogen in immunological experiments such as establishing animal models for hypersensitivity asthma etc EZ Cap OVA mRNA is provided at 1 mg/mL capped by co-transcriptional capping with a Cap 1 structure Compared to Cap 0 structures (such as ARCA and mCap) Cap 1-containing mRNA is more suitable for mammalian systems and exhibits higher transcription efficiency Additionally OVA mRNA is polyadenylated reducing host cell immune responses and enhancing mRNA stability and translational efficiency