Learn More
Apexbio Technology LLC EZ Cap OVA mRNA (m1Ψ) 1mg (1mg/mL)
SDP
Catalog No. 503107504
Encompass_Preferred
Ovalbumin (OVA) mRNA encodes a egg white glycoprotein with a molecular weight of approximately 45 000 daltons It can induce moderate immunogenicity in organisms and is commonly used as an immunogen in immunological experiments such as establishing animal models for hypersensitivity asthma etc EZ Cap OVA mRNA (m1 ) is provided at 1 mg/mL capped by co-transcriptional capping with a Cap 1 structure and polyadenylated Modified m1 incorporation reduces the host cell immune response and enhances mRNA stability and translation efficiency
Catalog No. 50-310-7504 Supplier Apexbio Technology LLC Supplier No. R10481
Ovalbumin (OVA) mRNA encodes a egg white glycoprotein with a molecular weight of approximately 45 000 daltons It can induce moderate immunogenicity in organisms and is commonly used as an immunogen in immunological experiments such as establishing animal models for hypersensitivity asthma etc EZ Cap OVA mRNA (m1 ) is provided at 1 mg/mL capped by co-transcriptional capping with a Cap 1 structure and polyadenylated Modified m1 incorporation reduces the host cell immune response and enhances mRNA stability and translation efficiency