Learn More
Brady™ EZ Panel Loc Snap-On Breaker Lockout
Catalog No. 19103955 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-955 Red
1 options
Catalog No. 19-103-955 Supplier Brady™ Supplier No. 51252
  • $49.50
    $38.65 / Each of 1
    Save $10.85
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Prevent switches from being thrown without proper justification. The Brady™ EZ Panel Loc Snap-On Breaker Lockout is as simple as aligning the pins with the hole and snapping closed.
  • The EZ Panel Loc System includes lockout devices, accomodating an exceptionally wide range of breaker models
  • The Snap-on breaker is for use on Square D NQO and other beakers with a hole in the switch tongue
  • Align pins with hole and snap closed
  • Pins prevent switch from being thrown
Specifications
Product Type Snap-on Circuit Breaker Lockout
Color Red
Material Fiberglass-reinforced Polyurethane
Description Breaker manufacturer: Square D
Dimensions (L x W x H) 0.9 x 1.88 x 0.74 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.