Learn More
Brady™ EZ Paneloc™ Circuit Breaker Lockout System Accessory - Spacing Gasket
Catalog No. 19103950 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
Catalog No. Color
19-103-950 White
1 options
Catalog No. 19-103-950 Supplier Brady™ Supplier No. 89256
Add to Cart
Add to Cart
For use with low profile panels, the EZ Paneloc™ spacing gasket pushes out the panel frame, adding an additional 3/8 in. of clearance between the panel and the door.
  • The EZ Paneloc™ spacing gasket allows low profile panel doors to be closed while EZ Paneloc™ lockouts are in place
  • Spacing gasket is easy to install with self-adhesive backing
  • Item includes spacing gasket only (EZ Paneloc™ lockout devices, lock rails and padlocks sold separately)
  • EZ Paneloc™ lock rail system allows panel door to be closed when lockouts are in place and fits multiple locks, eliminating the need for group hasps
Specifications
Product Type Spacing Gasket
Color White
Quantity 15 ft./Roll, 1 Roll/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.