Learn More
Brady™ EZ Paneloc™ Circuit Breaker Lockout System Components - Lock Rail
Catalog No. 19103947 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-103-947 Yellow
1 options
Catalog No. 19-103-947 Supplier Brady™ Supplier No. 65814
  • $65.25
    $39.65 / Pack of 1
    Save $25.60
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This lock rail is part of the EZ Paneloc™ lock rail system, which allows workers to secure EZ Paneloc™ circuit breaker lockouts by passing a plastic key through the lock rail.
  • EZ Paneloc™ lock rail system works on a wide range of breakers, including single- and multi-pole configurations
  • Lock rail works with most padlocks and permanently affixes to any location on the panel
  • Padlocks and EZ Paneloc™ Snap-On and Clamp-On breaker lockout devices sold separately
  • EZ Paneloc™ lock rail system allows panel door to be closed when lockouts are in place and fits multiple locks, eliminating the need for group hasps
Specifications
Product Type Lock Rail
Color Yellow
Quantity 6 Pk.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.