Learn More
Apexbio Technology LLC EZ Cap PEX-EGFP Probe mRNA (m1Ψ) 5x1mg (1mg/mL)
SDP
Catalog No. 503107398
Encompass_Preferred
PEX-EGFP mRNA encodes a fusion protein consisting of a peroxisome-targeting signal sequence fused to Enhanced Green Fluorescent Protein (EGFP) By fusing EGFP with the peroxisome targeting sequence researchers can observe the dynamic behavior distribution and function of peroxisomes in real-time in living cells aiding in revealing critical roles of peroxisomes in signaling pathways cellular metabolism and related diseases The EZ Cap PEX-EGFP Probe mRNA (m1 ) is provided at 1 mg/mL and is specifically designed for expressing fluorescent proteins targeted explicitly to peroxisomes This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The incorporation of m1 modifications poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo
Catalog No. 50-310-7398 Supplier Apexbio Technology LLC Supplier No. R11175
PEX-EGFP mRNA encodes a fusion protein consisting of a peroxisome-targeting signal sequence fused to Enhanced Green Fluorescent Protein (EGFP) By fusing EGFP with the peroxisome targeting sequence researchers can observe the dynamic behavior distribution and function of peroxisomes in real-time in living cells aiding in revealing critical roles of peroxisomes in signaling pathways cellular metabolism and related diseases The EZ Cap PEX-EGFP Probe mRNA (m1 ) is provided at 1 mg/mL and is specifically designed for expressing fluorescent proteins targeted explicitly to peroxisomes This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The incorporation of m1 modifications poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo