Learn More
Research Products International Corp EZ Pierce Adhesive Sealing Film, Non-Printed, 100 per Package
SDP
Catalog No. 50998789
Encompass
These adhesive sealing films provide the most convenient and positive method for sealing 96 well assay plates. Thermostable films form a superior seal around each well to prevent sample evaporation and cross contamination. Effective on both flat or raised rim plates. A split backing with removable end tabs allows fast and accurate positoning on plates. For sample recovery simply pierce film using common pipette tips or robotic metal probes. Adhesive is resistant to chemicals and solvents. EZ Pierce is a Trademark of Excel Scientific, Inc.
Catalog No. 50-998-789 Supplier Research Products International Corp Supplier No. 202519
These adhesive sealing films provide the most convenient and positive method for sealing 96 well assay plates. Thermostable films form a superior seal around each well to prevent sample evaporation and cross contamination. Effective on both flat or raised rim plates. A split backing with removable end tabs allows fast and accurate positoning on plates. For sample recovery simply pierce film using common pipette tips or robotic metal probes. Adhesive is resistant to chemicals and solvents. EZ Pierce is a Trademark of Excel Scientific, Inc.