Learn More
Thomas Scientific EZ-Pierce Sealing Film, Non-Sterile
SDP
Catalog No. NC1720448
Encompass
Easily pierceable, 70 µm thick, polyethylene with acrylic adhesive, EZ-Pierce™ sealing films allow direct sample recovery with single or multichannel pipettors and robotic probes. Dimensions are 82.6 by 142.9 mm for 96-well plates.
Catalog No. NC1720448 Supplier Thomas Scientific Supplier No. 6980A45
Easily pierceable, 70 µm thick, polyethylene with acrylic adhesive, EZ-Pierce™ sealing films allow direct sample recovery with single or multichannel pipettors and robotic probes. Dimensions are 82.6 by 142.9 mm for 96-well plates.