Learn More
Sigma Aldrich Fine Chemicals Biosciences EZ-PierceTM films sterile
Catalog No. 502900980
Encompass
Easily pierceable 70 mum thick polyethylene with acrylic adhesive EZ-Pierce films allow direct sample recovery with single or multichannel pipettors and robotic probes. Dimensions are 82.6 mm by 142.9 mm for 96 well plates.
Catalog No. 50-290-0980 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z72160350EA
Easily pierceable 70 mum thick polyethylene with acrylic adhesive EZ-Pierce films allow direct sample recovery with single or multichannel pipettors and robotic probes. Dimensions are 82.6 mm by 142.9 mm for 96 well plates.