Learn More
Alkali Scientific EZ-Punch™ Sealing Film, 100/pk
SDP
Catalog No. 502388714
Encompass_Preferred
EZ-Punch™ Sealing Film is designed for short-term processing and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes. Ideal for COVID-19 sample collection in deep-well plates using swabs. This pierceable film has printed circles that aid in the alignment of the film to the plate. Printed alphanumerics identify each well of a 96-well plate. Reseals after piercing for continued sample protection. To help position the film to the plate, align wells A-1 and H-12.
Catalog No. 50-238-8714 Supplier Alkali Scientific Supplier No. SF14ML
EZ-Punch™ Sealing Film is designed for short-term processing and storage of samples that require multiple access to wells by pipet tips or robotic probes. A pre-cut “X” over each well creates bendable flaps that provide sample access without coring or adhesive fouling of tips or probes. Ideal for COVID-19 sample collection in deep-well plates using swabs. This pierceable film has printed circles that aid in the alignment of the film to the plate. Printed alphanumerics identify each well of a 96-well plate. Reseals after piercing for continued sample protection. To help position the film to the plate, align wells A-1 and H-12.