Learn More
LAMDA BIOTECH INC EZ-Red Protein Blot Staining
Catalog No. NC3724892
Encompass_Preferred
EZ-Red™ is a specialized Ponceau S-based staining solution engineered for rapid and reversible protein visualization on Western blot membranes. This ready-to-use formulation enables quick assessment of protein transfer efficiency.
Catalog No. NC3724892 Supplier LAMDA BIOTECH INC Supplier No. GPS003
EZ-Red™ is a specialized Ponceau S-based staining solution engineered for rapid and reversible protein visualization on Western blot membranes. This ready-to-use formulation enables quick assessment of protein transfer efficiency.