Learn More
Zymo Research Corporation EZ RNA Methylation Kit (200 prep)
SDP
Catalog No. 502037281
Encompass_Preferred
The EZ RNA Methylation Kit features rapid and reliable bisulfite conversion of cytosines in RNA for methylation analysis. The kit streamlines the three-step process for complete conversion of cytosine in into uracil. RNA denaturation and bisulfite conversion processes are combined into a single step. No buffer preparation is necessary. The RNA Conversion Reagent is provided ready-to-use: simply add the reagent to an RNA sample and incubate as indicated. The innovative in-column desulphonation technology eliminates messy precipitation steps, ensuring researchers obtain consistent results. The product has been designed to minimize template degradation, loss of RNA during treatment and clean-up, and to provide complete conversion of cytosine for accurate methylation analysis. Recovered RNA is ideal for RT-PCR, sequencing, library preparation and Next-Gen sequencing.
Catalog No. 50-203-7281 Supplier Zymo Research Corporation Supplier No. R5002
The EZ RNA Methylation Kit features rapid and reliable bisulfite conversion of cytosines in RNA for methylation analysis. The kit streamlines the three-step process for complete conversion of cytosine in into uracil. RNA denaturation and bisulfite conversion processes are combined into a single step. No buffer preparation is necessary. The RNA Conversion Reagent is provided ready-to-use: simply add the reagent to an RNA sample and incubate as indicated. The innovative in-column desulphonation technology eliminates messy precipitation steps, ensuring researchers obtain consistent results. The product has been designed to minimize template degradation, loss of RNA during treatment and clean-up, and to provide complete conversion of cytosine for accurate methylation analysis. Recovered RNA is ideal for RT-PCR, sequencing, library preparation and Next-Gen sequencing.