Learn More
STEMCELL Technologies EasySep™ Direct Human Total Lymphocyte Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC1332746
Encompass
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications.​
Catalog No. NC1332746 Supplier STEMCELL Technologies Supplier No. 19655
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications.​