Learn More
STEMCELL Technologies EasySep™ Direct Human Pan-Granulocyte Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC1608088
Encompass
The EasySep™ Direct Human Pan-Granulocyte Isolation Kit isolates functional, highly purified neutrophils, eosinophils, and basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.
This item is not returnable. View return policy
Catalog No. NC1608088 Supplier STEMCELL Technologies Supplier No. 19659
The EasySep™ Direct Human Pan-Granulocyte Isolation Kit isolates functional, highly purified neutrophils, eosinophils, and basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.