Learn More
Paul N Gardner Company EZ SERIES VISCOSITY CUP NO3
SDP
Catalog No. NC0751694
Encompass
#3 Calibrated EZZAHN (ASTM) DIP CUP produced in accordance with ISO/IEC 17025:2005, ISO/IEC Guide 34:2009, ISO 9001:2008; are compatible to ASTM D 1084 Viscosity of Adhesives and ASTM D 816; calibration is traceable to NIST
Catalog No. NC0751694 Supplier Paul N Gardner Company Supplier No. EZ3
#3 Calibrated EZZAHN (ASTM) DIP CUP produced in accordance with ISO/IEC 17025:2005, ISO/IEC Guide 34:2009, ISO 9001:2008; are compatible to ASTM D 1084 Viscosity of Adhesives and ASTM D 816; calibration is traceable to NIST