Learn More
Braintree Scientific EZ Sure-Seal Chamber Guinea Pig/Rabbit 12"" x 18 x 12
SDP
Catalog No. 501954783
Encompass
Sure-Seal Induction Chambers. Initial anesthesia is performed in the induction chamber before animals are moved to the surgical bed for procedure. Positive seal gasket ensures there is no gas leakage. Simple locking snap latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Can be oxygen-flushed before transferring animals out of chamber. Constructed of heavy-duty, clear acrylic plastic. Non-skid bottom **Sold for research purposes only and not for human use**
Catalog No. 50-195-4783 Supplier Braintree Scientific Supplier No. EZ179
Sure-Seal Induction Chambers. Initial anesthesia is performed in the induction chamber before animals are moved to the surgical bed for procedure. Positive seal gasket ensures there is no gas leakage. Simple locking snap latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Can be oxygen-flushed before transferring animals out of chamber. Constructed of heavy-duty, clear acrylic plastic. Non-skid bottom **Sold for research purposes only and not for human use**