Learn More
Cayman Chemical EZ-T-603 1mg
SDP
Catalog No. 502916689
Encompass_Preferred
A mixture of (E)-IT-603 and (Z)-IT-603; (Z)-IT-603 is a c-Rel inhibitor (IC50 = 3 µM); (Z)-IT-603 inhibits c-Rel DNA binding in an EMSA; preincubation with mouse T cells prevents the development of GVHD but does not affect GVT activity in a mouse model of allogeneic hematopoietic stem cell transplantation at 20 µM
Catalog No. 50-291-6689 Supplier Cayman Chemical Supplier No. 377921 MG
A mixture of (E)-IT-603 and (Z)-IT-603; (Z)-IT-603 is a c-Rel inhibitor (IC50 = 3 µM); (Z)-IT-603 inhibits c-Rel DNA binding in an EMSA; preincubation with mouse T cells prevents the development of GVHD but does not affect GVT activity in a mouse model of allogeneic hematopoietic stem cell transplantation at 20 µM