Learn More
Julabo USA Inc Each, EasyTemp Professional control software
SDP
Catalog No. 507534275
Encompass_Preferred
Bath Accessories, Julabo, EasyTemp Professional control software. Enables control of up to 24 JULABO devices with data recording, ramp programming, formula implementation and loops.
Catalog No. 50-753-4275 Supplier Julabo USA Inc Supplier No. 8901105
Bath Accessories, Julabo, EasyTemp Professional control software. Enables control of up to 24 JULABO devices with data recording, ramp programming, formula implementation and loops.