Learn More
MESA LABS BOZEMAN MFG FACILITY SMART-READ STEAM log 5 100ct
SDP
Catalog No. NC0566228
Encompass
SMART-READ EZ TEST. The Smart-Read EZTest Biological Monitoring System allows an organization to release sterile product with true biological confirmation faster and easier than ever before. This unique system uses a BI – with no added enzyme – which is incubated, evaluated and documented in one simple, automated operation. Relying only upon bacterial spore growth, the Smart-Read system can detect sterilization failure in as few as three to five hours, and negative results can be confirmed with ten hours of incubation. G. stearothermophilus - 7953. Population: 10 log 5. Packaging: 100 count.
This item is not returnable. View return policy
Catalog No. NC0566228 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. SEZS/5
SMART-READ EZ TEST. The Smart-Read EZTest Biological Monitoring System allows an organization to release sterile product with true biological confirmation faster and easier than ever before. This unique system uses a BI – with no added enzyme – which is incubated, evaluated and documented in one simple, automated operation. Relying only upon bacterial spore growth, the Smart-Read system can detect sterilization failure in as few as three to five hours, and negative results can be confirmed with ten hours of incubation. G. stearothermophilus - 7953. Population: 10 log 5. Packaging: 100 count.