Learn More
MESA LABS BOZEMAN MFG FACILITY EZ TEST STEAM 25/PK
SDP
Catalog No. NC9493657
Encompass
EZ-Test 10e5 BI's for steam sterilization 25/box
Catalog No. NC9493657 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. EZS/525
EZ-Test 10e5 BI's for steam sterilization 25/box