Learn More
MESA LABS BOZEMAN MFG FACILITY EZ TST BIO INDICATORS 100/BX
SDP
Catalog No. NC9973359
Encompass
EZTest Self-Contained Color Change Indicators, For Steam And Ethylene Oxide Gas, biological indicator for monitoring steam or ethylene oxide sterilization
Catalog No. NC9973359 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. EZS/6
EZTest Self-Contained Color Change Indicators, For Steam And Ethylene Oxide Gas, biological indicator for monitoring steam or ethylene oxide sterilization