Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories EZ Vitamin D (25-OH) Assay Liquid Kit, 2-reagent, liquid stable, 200 Tests, 3 μL sample volume, 1 x 40 mL R1, 1 x 10 mL R2, 10 min time to results

Catalog No. NC2333613
Encompass_Preferred

The Diazyme EZ Vitamin D Assay is designed for the quantitative determination of 25-hydroxyvitamin D (25-OH vitamin D) in human serum and plasma using automated clinical chemistry analyzers. This assay provides a precise and efficient solution for assessing vitamin D sufficiency by detecting both Vitamin D2 and D3 without requiring sample pre-treatment or pre-dilution. It features high throughput capabilities and is traceable to NIST standards. With FDA clearance and CE marking, it ensures compliance and reliability in clinical settings.

  • Liquid, two-reagent format
  • 200 tests per kit
  • Sample volume of 3 μL
  • Time to results of approximately 10 min
  • Throughput of approximately 300 tests/hour
  • Linear range of 7.6 - 147.8 ng/mL
  • Precision of <10% coefficient of variation
  • Regulatory certified by FDA and CE
  • For use with automated clinical chemistry analyzers

Catalog No. NC2333613 Supplier Diazyme Laboratories Supplier No. DZ888AK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Diazyme EZ Vitamin D Assay is designed for the quantitative determination of 25-hydroxyvitamin D (25-OH vitamin D) in human serum and plasma using automated clinical chemistry analyzers. This assay provides a precise and efficient solution for assessing vitamin D sufficiency by detecting both Vitamin D2 and D3 without requiring sample pre-treatment or pre-dilution. It features high throughput capabilities and is traceable to NIST standards. With FDA clearance and CE marking, it ensures compliance and reliability in clinical settings.

  • Liquid, two-reagent format
  • 200 tests per kit
  • Sample volume of 3 μL
  • Time to results of approximately 10 min
  • Throughput of approximately 300 tests/hour
  • Linear range of 7.6 - 147.8 ng/mL
  • Precision of <10% coefficient of variation
  • Regulatory certified by FDA and CE
  • For use with automated clinical chemistry analyzers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.