Learn More
Medicus Health EZ2 Freeze Hard Side Ice Packs, Green, Medium, 1.5 lbs.
SDP
Catalog No. NC4027746
Encompass
• EZ2 Freeze Hard Side Specimen Transport Ice Packs, Medium, Green, 1.5 lbs., 6.75"L x 7.75" Wx 1.25"D
• Specimen Transport Ice Packs freeze at 32°F, efficiently keeping specimens cold without the risk of freezing and the hassle and mess of ice
• Integrated handles make these ice packs easy to transport and rugged rigid body make the easy to stack
• Tested to maintain food-safe temperatures for hours, even days without the risk of freezing
• When finished using, simply store in freezer until needed again
• Freeze the ice panels in any freezer 6-8 hours prior to use
Catalog No. NC4027746 Supplier Medicus Health Supplier No. 5307M18
• EZ2 Freeze Hard Side Specimen Transport Ice Packs, Medium, Green, 1.5 lbs., 6.75"L x 7.75" Wx 1.25"D
• Specimen Transport Ice Packs freeze at 32°F, efficiently keeping specimens cold without the risk of freezing and the hassle and mess of ice
• Integrated handles make these ice packs easy to transport and rugged rigid body make the easy to stack
• Tested to maintain food-safe temperatures for hours, even days without the risk of freezing
• When finished using, simply store in freezer until needed again
• Freeze the ice panels in any freezer 6-8 hours prior to use