Learn More
Zymo Research Corporation EZ-96 DNA Methlyation-Direct™ Kit (Deep-Well) (2 x 96 Rxns)
SDP
Catalog No. 50444333
Encompass_Preferred
The increased sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50 pg DNA. Like the EZ DNA Methylation-Gold™ Kit, DNA denaturation and bisulfite conversion processes are combined into a single step. All kits streamline the three step process of bisulfite conversion of nonmethylated cytosine in DNA into uracil. In addition the methylation kits share innovative in-column desulphonation technology that eliminates otherwise cumbersome DNA precipitation steps while ensuring researchers consistent results every time. The kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for PCR amplification for downstream analyses including restriction endonuclease digestion, sequencing, microarrays, etc.
This item is not returnable. View return policy
Catalog No. 50-444-333 Supplier Zymo Research Corporation Supplier No. D5023
Add to Cart
Add to Cart
The increased sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50 pg DNA. Like the EZ DNA Methylation-Gold™ Kit, DNA denaturation and bisulfite conversion processes are combined into a single step. All kits streamline the three step process of bisulfite conversion of nonmethylated cytosine in DNA into uracil. In addition the methylation kits share innovative in-column desulphonation technology that eliminates otherwise cumbersome DNA precipitation steps while ensuring researchers consistent results every time. The kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for PCR amplification for downstream analyses including restriction endonuclease digestion, sequencing, microarrays, etc.