Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Lightning Kit (Deep-Well) (2x96 rxns.)
SDP
Catalog No. 50444342
Encompass_Preferred
The EZ-96 DNA Methylation-Lightning Kit is a rapid, high-throughput (96-well) kit for DNA methylation analysis. The streamlined workflow features ready-to-use Lightning Conversion Reagent and a combined DNA denaturation and bisulfite conversion process. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. With the Deep-Well Kit (Cat. D5033), samples can be eluted in as little as 15 l. High yield, converted DNA is ideal for PCR, array, and Next-Generation sequencing, etc.
Catalog No. 50-444-342 Supplier Zymo Research Corporation Supplier No. D5033
The EZ-96 DNA Methylation-Lightning Kit is a rapid, high-throughput (96-well) kit for DNA methylation analysis. The streamlined workflow features ready-to-use Lightning Conversion Reagent and a combined DNA denaturation and bisulfite conversion process. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. With the Deep-Well Kit (Cat. D5033), samples can be eluted in as little as 15 l. High yield, converted DNA is ideal for PCR, array, and Next-Generation sequencing, etc.