Learn More
Zymo Research Corporation EZ-96 DNA Methylation™ Kit (Deep-Well) (2 x 96 Rxns)
SDP
Catalog No. 50444293
Encompass_Preferred
The EZ-96 DNA Methylation Kits features a high-throughput (96-well spin-plate), simplified procedure that streamlines bisulfite conversion of DNA. Cytosines undergo a three-step reaction with sodium bisulfite during which the cytosine is converted into uracil. The innovative in-column desulphonation technology eliminate otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc. Note: Catalog # D5004 is recommended for use with the Illumina Infinium MethylationEPIC BeadChip array.
Catalog No. 50-444-293 Supplier Zymo Research Corporation Supplier No. D5004
The EZ-96 DNA Methylation Kits features a high-throughput (96-well spin-plate), simplified procedure that streamlines bisulfite conversion of DNA. Cytosines undergo a three-step reaction with sodium bisulfite during which the cytosine is converted into uracil. The innovative in-column desulphonation technology eliminate otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc. Note: Catalog # D5004 is recommended for use with the Illumina Infinium MethylationEPIC BeadChip array.