Learn More
Foxx Life Sciences EZBio 115L (30 Gal) Carboy, HDPE, Wide Mouth 120mm VersaCap, 120L Max Capacity, Double Bagged, Single Use Space Saving Rectangular Carboy with Handles and Graduation Marks , Sterile, 1/EA
Catalog No. 5000078003
Encompass_Preferred
Foxx Life Sciences EZBio Carboy is a versatile fluid handling system designed for efficiency and ease of use. Made from USP Class VI, FDA-grade materials, it’s available sterile or non-sterile.
The rectangular shape saves bench space, and the wide-mouth design allows easy filling and cleaning. Its ergonomic body includes top handles and molded grips for safe handling. Each carboy features VersaCap Technology with leak-proof, interchangeable top connections and adapter inserts. Graduations are certified to ±5% accuracy.Designed for bioprocessing and fluid storage. Also functions as a lab fluid handling container or a laboratory carboy
Features
Gamma sterilized, cleanroom double-bagged (sterile models)
Ergonomic design with molded grips
Wide-mouth for easy use
Certified graduation marks
Applications
Bioprocess container for media formulation and mixing
Aseptic processes
Handling aggressive chemicals
Production with low leachables/extractables
Catalog No. 50-000-78003 Supplier Foxx Life Sciences Supplier No. 5125M61FLS
Foxx Life Sciences EZBio Carboy is a versatile fluid handling system designed for efficiency and ease of use. Made from USP Class VI, FDA-grade materials, it’s available sterile or non-sterile.
The rectangular shape saves bench space, and the wide-mouth design allows easy filling and cleaning. Its ergonomic body includes top handles and molded grips for safe handling. Each carboy features VersaCap Technology with leak-proof, interchangeable top connections and adapter inserts. Graduations are certified to ±5% accuracy.Designed for bioprocessing and fluid storage. Also functions as a lab fluid handling container or a laboratory carboy
Features
Gamma sterilized, cleanroom double-bagged (sterile models)
Ergonomic design with molded grips
Wide-mouth for easy use
Certified graduation marks
Applications
Bioprocess container for media formulation and mixing
Aseptic processes
Handling aggressive chemicals
Production with low leachables/extractables