Learn More
Foxx Life Sciences EZBio 115L (30 Gal) Carboy, Polypropylene (PP), Autoclavable, Wide Mouth 120mm VersaCap, 120L Max Capacity, Double Bagged, Single Use Space Saving Rectangular Carboy with Handles and Graduation Marks, Sterile, 1/EA
Catalog No. 5000078000
Encompass_Preferred
EZBio Carboys are versatile, autoclavable lab containers designed for cleanroom or laboratory use. The rectangular shape saves bench space, while large handles and molded base grips provide a secure, ergonomic hold for safe lifting and pouring. Made from durable PP, each carboy features a wide-mouth opening for easy cleaning and filling. Volume and material type are clearly marked, with large, certified graduations accurate to ±5%.
Features
PP carboy with handles for ergonomic use
Leak-proof carboy cap system (VersaCap) with interchangeable connections
EZfil™ wide-mouth for easy access
VersaBarb allows spigot flexibility
Autoclavable at 121°C for 20 min at 15 PSI
Large-volume lab container with certified graduations
Applications
Bioprocessing container for liquid or powder storage
Autoclave sterilization
Bulk dispensing (e.g., APIs)
Heavy-duty or vacuum use
Catalog No. 50-000-78000 Supplier Foxx Life Sciences Supplier No. 5155M61FLS
EZBio Carboys are versatile, autoclavable lab containers designed for cleanroom or laboratory use. The rectangular shape saves bench space, while large handles and molded base grips provide a secure, ergonomic hold for safe lifting and pouring. Made from durable PP, each carboy features a wide-mouth opening for easy cleaning and filling. Volume and material type are clearly marked, with large, certified graduations accurate to ±5%.
Features
PP carboy with handles for ergonomic use
Leak-proof carboy cap system (VersaCap) with interchangeable connections
EZfil™ wide-mouth for easy access
VersaBarb allows spigot flexibility
Autoclavable at 121°C for 20 min at 15 PSI
Large-volume lab container with certified graduations
Applications
Bioprocessing container for liquid or powder storage
Autoclave sterilization
Bulk dispensing (e.g., APIs)
Heavy-duty or vacuum use