Learn More
Foxx Life Sciences 1L HDPE Media Bottle, GL45 Cap, Sterile, 12/PK
GREENER_CHOICE
Catalog No. 5000555608
Encompass_Preferred
EZBiopure S 1L HDPE Square Storage Bottles with graduation marks, GL45 Cap, Bleach Compatible, Individually Double Bagged ideal for SUT bioprocess applications. The 1L square HDPE media bottles offers durable, chemical-resistant storage for reagents, buffers, and media.
Features
Chemical resistant media Bottles HDPE material for safe storage
Square profile design for space-efficient storage and improved shelf utilization
Narrow Mouth HDPE bottles with GL45 VersaCap
Made entirely of 100% USP Class VI Components
HDPE Clear Media Bottles have graduation marks for easy volume measurement and reference
The Square HDPE Bottle are manufactured in a Class 7 cleanroom at an ISO 13485–certified FOXX facility.
Applications
Reagent storage
Media and buffer preparation
Feed and intermediate solution handling
Catalog No. 50-005-55608 Supplier Foxx Life Sciences Supplier No. 5122A33FLS
EZBiopure S 1L HDPE Square Storage Bottles with graduation marks, GL45 Cap, Bleach Compatible, Individually Double Bagged ideal for SUT bioprocess applications. The 1L square HDPE media bottles offers durable, chemical-resistant storage for reagents, buffers, and media.
Features
Chemical resistant media Bottles HDPE material for safe storage
Square profile design for space-efficient storage and improved shelf utilization
Narrow Mouth HDPE bottles with GL45 VersaCap
Made entirely of 100% USP Class VI Components
HDPE Clear Media Bottles have graduation marks for easy volume measurement and reference
The Square HDPE Bottle are manufactured in a Class 7 cleanroom at an ISO 13485–certified FOXX facility.
Applications
Reagent storage
Media and buffer preparation
Feed and intermediate solution handling