Learn More
Foxx Life Sciences 1L PP Media Bottle, GL45 Cap, Non Sterile, 12/PK
GREENER_CHOICE
Catalog No. 5000555600
Encompass_Preferred
EZBiopure S 1L PP Square Storage Bottles with graduation marks, GL45 Cap, Autoclavable, Individually Double Bagged ideal for SUT bioprocess applications. The 1L square PP media bottles offers durable, chemical-resistant storage for reagents, buffers, and media.
Features
Chemical resistant media Bottles PP material for safe storage
Square profile design for space-efficient storage and improved shelf utilization
Narrow Mouth PP bottles with GL45 VersaCap
Made entirely of 100% USP Class VI Components
PP Clear Media Bottles have graduation marks for easy volume measurement and reference
The Square PP Bottle are manufactured in a Class 7 cleanroom at an ISO 13485–certified FOXX facility.
Applications
Reagent storage
Media and buffer preparation
Feed and intermediate solution handling
Catalog No. 50-005-55600 Supplier Foxx Life Sciences Supplier No. 5152A31FLS
EZBiopure S 1L PP Square Storage Bottles with graduation marks, GL45 Cap, Autoclavable, Individually Double Bagged ideal for SUT bioprocess applications. The 1L square PP media bottles offers durable, chemical-resistant storage for reagents, buffers, and media.
Features
Chemical resistant media Bottles PP material for safe storage
Square profile design for space-efficient storage and improved shelf utilization
Narrow Mouth PP bottles with GL45 VersaCap
Made entirely of 100% USP Class VI Components
PP Clear Media Bottles have graduation marks for easy volume measurement and reference
The Square PP Bottle are manufactured in a Class 7 cleanroom at an ISO 13485–certified FOXX facility.
Applications
Reagent storage
Media and buffer preparation
Feed and intermediate solution handling