Learn More
Foxx Life Sciences Foxx Life Sciences’ EZBio Carboy
Catalog No. NC2311534
Encompass_Preferred
Foxx Life Sciences’ EZBio Carboy is an advanced fluid handling system developed with the end-user in mind. The EZBio Carboy serves as a highly versatile container, suitable for all fluid management applications. Assemblies are made entirely of USP Class VI, FDA Grade materials and are available Sterile or Non-Sterile, double bagged
Catalog No. NC2311534 Supplier Foxx Life Sciences Supplier No. 5175G51FLS
Foxx Life Sciences’ EZBio Carboy is an advanced fluid handling system developed with the end-user in mind. The EZBio Carboy serves as a highly versatile container, suitable for all fluid management applications. Assemblies are made entirely of USP Class VI, FDA Grade materials and are available Sterile or Non-Sterile, double bagged