Learn More
Foxx Life Sciences EZBio Rectangular Carboy with 6" TC Sanitary Neck, No Spigot, Polypropylene (PP), 20L Carboy with Handle, Powder Carboy, Autoclavable, Sterile, 1/CS
Catalog No. 5000304244
Encompass_Preferred
EZBio Carboy with Sanitary Neck is a fluid and powder handling container designed for bioprocessing, laboratory, and pharmaceutical applications. Designed for closed-system fluid, it features 4” and 6” tri-clamp sanitary necks, ensuring contamination-free connections in cleanroom environments. Available in round and rectangular formats, the round sanitary carboy supports efficient mixing, while the rectangular design provides space-saving storage. Constructed from high-purity plastic this tri-clamp carboy delivers reliable performance for bioprocess production, buffer and media preparation, sampling, and waste collection—making it a trusted solution across upstream and downstream workflows.
Features
4” and 6” tri-clamp sanitary necks for closed-loop operation
Materials comply with USP Class VI standards
BPA-free, gamma-sterilizable
Wide-mouth ergonomic design for easy filling and cleaning
Ideal for buffer/media prep, powder transfer, and waste collection
Catalog No. 50-003-04244 Supplier Foxx Life Sciences Supplier No. 5155H91FLS
EZBio Carboy with Sanitary Neck is a fluid and powder handling container designed for bioprocessing, laboratory, and pharmaceutical applications. Designed for closed-system fluid, it features 4” and 6” tri-clamp sanitary necks, ensuring contamination-free connections in cleanroom environments. Available in round and rectangular formats, the round sanitary carboy supports efficient mixing, while the rectangular design provides space-saving storage. Constructed from high-purity plastic this tri-clamp carboy delivers reliable performance for bioprocess production, buffer and media preparation, sampling, and waste collection—making it a trusted solution across upstream and downstream workflows.
Features
4” and 6” tri-clamp sanitary necks for closed-loop operation
Materials comply with USP Class VI standards
BPA-free, gamma-sterilizable
Wide-mouth ergonomic design for easy filling and cleaning
Ideal for buffer/media prep, powder transfer, and waste collection